accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79236-92-3

IUPAC Name: 1,4,7,10-tetraazatetracyclo[5.5.2.0⁴,¹³.0¹⁰,¹⁴]tetradecane
Synonyms: 1,4,7,10-tetraazatetracyclo[5.5.2.0⁴,¹³.0¹⁰,¹⁴]tetradecane
Molecular Weight (g/mol): 194.28
Molecular Formula: C10H18N4
InChi Key: YSPZOYMEWUTYDA-UHFFFAOYSA-N
SMILES: C1CN2CCN3CCN4CCN1C2C34
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results