CAS RN 79338-14-0
IUPAC Name:
4-amino-5-(benzyloxy)-5-oxopentanoic acid
Synonyms:
4-amino-5-(benzyloxy)-5-oxopentanoic acid
Molecular Weight (g/mol):
237.26
Molecular Formula:
C12H15NO4
InChi Key:
HFZKKJHBHCZXTQ-UHFFFAOYNA-N
SMILES:
NC(CCC(O)=O)C(=O)OCC1=CC=CC=C1