accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79538-29-7

FFFClO

CAS RN 79538-29-7

IUPAC Name: 2,4,6-trifluorobenzoyl chloride
Synonyms: 2,4,6-trifluorobenzoyl chloride
Molecular Weight (g/mol): 194.54
Molecular Formula: C7H2ClF3O
InChi Key: SIFIJQFBERMWMU-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C(Cl)=O)C(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results