accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79583-19-0

CH3NNOCl

CAS RN 79583-19-0

IUPAC Name: 1-methyl-1H-pyrazole-4-carbonyl chloride
Synonyms: 1-methylpyrazole-4-carbonyl chloride
Molecular Weight (g/mol): 144.56
Molecular Formula: C5H5ClN2O
InChi Key: QLBBQLJPRXPVOS-UHFFFAOYSA-N
SMILES: CN1C=C(C=N1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results