CAS RN 79617-98-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R,R)-Sertraline, TRC
CAS: 79617-98-4 Molecular Formula: C17 H17 Cl2 N Molecular Weight (g/mol): 306.2296 Synonym: (1R,4R)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine,(1R-cis)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-Naphthalenamine,(1R,4R)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine,CP 51973 IUPAC Name: (1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine SMILES: CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13