accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79723-02-7

CH3NCH3CH3H3COHOOO

CAS RN 79723-02-7

IUPAC Name: tetramethylazanium 2-carboxybenzoate
Synonyms: hydrogen phthalate; tetramethylammonium ion
Molecular Weight (g/mol): 239.27
Molecular Formula: C12H17NO4
InChi Key: KQTFRASJMFIJPC-UHFFFAOYSA-M
SMILES: C[N+](C)(C)C.OC(=O)C1=CC=CC=C1C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results