accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79784-34-2

IUPAC Name: 2-(3,4-difluorophenyl)-2-oxoacetaldehyde
Synonyms: 2-(3,4-difluorophenyl)-2-oxoacetaldehyde
Molecular Weight (g/mol): 170.12
Molecular Formula: C8H4F2O2
InChi Key: VZRYIZGMQICGSF-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1F)C(=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results