accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOO

CAS RN 79814-40-7

IUPAC Name: 2,5-dioxooxolan-3-yl acetate
Synonyms: 2,5-dioxooxolan-3-yl acetate
Molecular Weight (g/mol): 158.11
Molecular Formula: C6H6O5
InChi Key: SSWJHSASZZAIAU-UHFFFAOYNA-N
SMILES: CC(=O)OC1CC(=O)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results