accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79887-09-5

IUPAC Name: 1-butyl-4-ethynylbenzene
Synonyms: 1-butyl-4-ethynylbenzene
Molecular Weight (g/mol): 158.24
Molecular Formula: C12H14
InChi Key: ZVWWYEHVIRMJIE-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results