accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79887-11-9

IUPAC Name: 1-ethynyl-4-hexylbenzene
Synonyms: 1-ethynyl-4-hexylbenzene
Molecular Weight (g/mol): 186.3
Molecular Formula: C14H18
InChi Key: NFPDFDTYANKKIU-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results