accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79887-16-4

H3COCH

CAS RN 79887-16-4

IUPAC Name: 1-ethynyl-4-(pentyloxy)benzene
Synonyms: 1-ethynyl-4-(pentyloxy)benzene
Molecular Weight (g/mol): 188.27
Molecular Formula: C13H16O
InChi Key: MKSWQHOPSDCVMS-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(C=C1)C#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)