accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNFHCl

CAS RN 799274-06-9

IUPAC Name: 7-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Synonyms: 7-fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Molecular Weight (g/mol): 187.64
Molecular Formula: C9H11ClFN
InChi Key: LOJNQXIJLCPQDR-UHFFFAOYSA-N
SMILES: Cl.FC1=CC2=C(CCNC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results