accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 799557-87-2

IUPAC Name: 2-chloro-5-(trifluoromethyl)pyrazine
Synonyms: 2-chloro-5-(trifluoromethyl)pyrazine
Molecular Weight (g/mol): 182.53
Molecular Formula: C5H2ClF3N2
InChi Key: AIEGIFIEQXZBCP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(Cl)C=N1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)