accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 79999-47-6

CH3H3CCH3SiH3CCH3OOOSiCH3H3CCH3CH3H3COSiCH3H3CCH3H3CCH3

CAS RN 79999-47-6

IUPAC Name: tert-butyl({4-[(tert-butyldimethylsilyl)oxy]-2-{[(tert-butyldimethylsilyl)oxy]methyl}-3,4-dihydro-2H-pyran-3-yl}oxy)dimethylsilane
Synonyms: tert-butyl({4-[(tert-butyldimethylsilyl)oxy]-2-{[(tert-butyldimethylsilyl)oxy]methyl}-3,4-dihydro-2H-pyran-3-yl}oxy)dimethylsilane
Molecular Weight (g/mol): 488.93
Molecular Formula: C24H52O4Si3
InChi Key: FMXIGEXRLSNRMR-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](C)(C)OCC1OC=CC(O[Si](C)(C)C(C)(C)C)C1O[Si](C)(C)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results