accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClSiCl

CAS RN 80-10-4

IUPAC Name: dichlorodiphenylsilane
Synonyms: dichlorodiphenylsilane
Molecular Weight (g/mol): 253.2
Molecular Formula: C12H10Cl2Si
InChi Key: OSXYHAQZDCICNX-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results