accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNHSOO

CAS RN 80-17-1

IUPAC Name: benzenesulfonohydrazide
Synonyms: benzenesulfonic acid hydrazide
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: VJRITMATACIYAF-UHFFFAOYSA-N
SMILES: NNS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results