accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 80-56-8

CH3H3CH3C

CAS RN 80-56-8

IUPAC Name: 2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Synonyms: α pinene fema number 2902 1R-a-pinene α-pinene (product) acintene A 1R-(+)-a-pinene 1R-α-pinene α-pinene (substance) wilt pruf pinene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: GRWFGVWFFZKLTI-UHFFFAOYNA-N
SMILES: CC1=CCC2CC1C2(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results