accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 80-70-6

IUPAC Name: N,N,N',N'-tetramethylguanidine
Synonyms: 1,1,3,3-tetramethylguanidine
Molecular Weight (g/mol): 115.18
Molecular Formula: C5H13N3
InChi Key: KYVBNYUBXIEUFW-UHFFFAOYSA-N
SMILES: CN(C)C(=N)N(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (4)