accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNH3CO

CAS RN 80-73-9

IUPAC Name: 1,3-dimethylimidazolidin-2-one
Synonyms: dimethyl imidazolidinone
Molecular Weight (g/mol): 114.15
Molecular Formula: C5H10N2O
InChi Key: CYSGHNMQYZDMIA-UHFFFAOYSA-N
SMILES: CN1CCN(C)C1=O
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (5)
Boiling Point 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)