accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 8000-41-7

IUPAC Name: 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
Synonyms: terpineol 1-α-terpineol α-terpineol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: WUOACPNHFRMFPN-UHFFFAOYNA-N
SMILES: CC1=CCC(CC1)C(C)(C)O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)