accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 8001-79-4

IUPAC Name: 1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
Synonyms: ricinus communis seed oil ricinus communis root extract
Molecular Weight (g/mol): 933.45
Molecular Formula: C57H104O9
InChi Key: ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
SMILES: CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)