accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 8004-13-5

IUPAC Name: 1,1'-biphenyl; phenoxybenzene
Synonyms: diphenyl; diphenyl-ether
Molecular Weight (g/mol): 324.42
Molecular Formula: C24H20O
InChi Key: MHCVCKDNQYMGEX-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=CC=C1.O(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)