CAS RN 80049-61-2
CAS RN 80049-61-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Promotions Available
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate, 98%
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate, 98%
CAS: 80049-61-2 Molecular Formula: C11H14F6N3PRu Molecular Weight (g/mol): 434.29 MDL Number: MFCD02684569
2
Tris(acetonitrile)cyclopentadienylruthenium(II) Hexafluorophosphate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tris(acetonitrile)cyclopentadienylruthenium(II) Hexafluorophosphate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Tris(acetonitrile)cyclopentadienylruthenium(II) Hexafluorophosphate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.