accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 80049-87-2

IUPAC Name: 9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms: 9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Molecular Weight (g/mol): 270.22
Molecular Formula: C10H11FN4O4
InChi Key: NRVOTDBYJXFINS-QYYRPYCUSA-N
SMILES: OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1NC=NC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results