accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 8006-28-8

IUPAC Name: sodium oxocalcium hydroxide
Synonyms: sodium calcium oxide hydroxide
Molecular Weight (g/mol): 96.07
Molecular Formula: CaHNaO2
InChi Key: HUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES: [OH-].[Na+].O=[Ca]
Molecular Weight (g/mol) 
  • (1)
  • (18)
Quantity 
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (7)
Grade 
  • (2)
  • (3)
  • (3)
  • (6)
Physical Form 
  • (4)
  • (7)
Material 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (9)
  • (2)
  • (1)
  • (18)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (7)
  • (2)
  • (3)
  • (3)
  • (6)
  • (4)
  • (7)
  • (8)