accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 8009-03-8

IUPAC Name: 1,1,2-trimethyl-1H-benzo[e]indole
Synonyms: petroleum jelly
Molecular Weight (g/mol): 209.29
Molecular Formula: C15H15N
InChi Key: WJZSZXCWMATYFX-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results