CAS RN 80109-63-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Desoxyquinocetone, TRC
CAS: 80109-63-3 Molecular Formula: C18H14N2O Molecular Weight (g/mol): 274.32 Synonym: 1-(3-Methyl-2-quinoxalinyl)-3-phenyl-2-propen-1-one; SMILES: CC1=NC2=CC=CC=C2N=C1C(=O)/C=C/C3=CC=CC=C3