accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 8012-95-1

IUPAC Name: 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-1λ⁴-chromen-1-ylium chloride
Synonyms: liquid paraffin (drug)
Molecular Weight (g/mol): 338.7
Molecular Formula: C15H11ClO7
InChi Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N
SMILES: [Cl-].OC1=CC2=[O+]C(=C(O)C=C2C(O)=C1)C1=CC(O)=C(O)C(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (4)
Grade 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)