CAS RN 8013-01-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Thermo Scientific Chemicals Yeast extract, for biochemistry
CAS: 8013-01-2
2
Yeast Extract, Powder, MP Biomedicals™
CAS: 8013-01-2