accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

MnZnSSNHHNSSSSNHHNSS

CAS RN 8018-01-7

IUPAC Name: manganese(2+) zinc(2+) bis(({2-[(sulfanidylmethanethioyl)amino]ethyl}carbamothioyl)sulfanide)
Synonyms: mancozeb mancozeb (iso)
Molecular Weight (g/mol): 541.01
Molecular Formula: C8H12MnN4S8Zn
InChi Key: CHNQZRKUZPNOOH-UHFFFAOYSA-J
SMILES: [Mn++].[Zn++].[S-]C(=S)NCCNC([S-])=S.[S-]C(=S)NCCNC([S-])=S
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results