accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHOFFFFFFFFFFFF

CAS RN 802-93-7

IUPAC Name: 1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
Synonyms: 1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
Molecular Weight (g/mol): 410.16
Molecular Formula: C12H6F12O2
InChi Key: PGUIOHNOYADLMU-UHFFFAOYSA-N
SMILES: OC(C1=CC(=CC=C1)C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)