accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOONOO

CAS RN 80307-12-6

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)butanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 4-(2,5-dioxopyrrol-1-yl)butanoate
Molecular Weight (g/mol): 280.24
Molecular Formula: C12H12N2O6
InChi Key: PVGATNRYUYNBHO-UHFFFAOYSA-N
SMILES: O=C(CCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)