accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3OHOH3C

CAS RN 8050-09-7

IUPAC Name: 1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
Synonyms: rosin tall oil rosin
Molecular Weight (g/mol): 302.46
Molecular Formula: C20H30O2
InChi Key: RSWGJHLUYNHPMX-UHFFFAOYNA-N
SMILES: CC(C)C1=CC2=CCC3C(C)(CCCC3(C)C(O)=O)C2CC1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)