accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 80565-30-6

IUPAC Name: 4'-chloro-[1,1'-biphenyl]-4-carbaldehyde
Synonyms: 4'-chloro-[1,1'-biphenyl]-4-carbaldehyde
Molecular Weight (g/mol): 216.66
Molecular Formula: C13H9ClO
InChi Key: UXCMNUUPBMYDLJ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC=C(C=O)C=C1
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results