accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3O(S)O(S)CH3OH3CHOHONHO(S)H3C(S)HO(R)H3C(R)HO(R)H3C(S)OH3CO(R)H3CCH3NCH3NO(?)(Z)(Z)

CAS RN 80621-81-4

IUPAC Name: (7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-2,15,17,36-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22,30-octamethyl-6,23-dioxo-8,37-dioxa-24,27,33-triazahexacyclo[23.10.1.1⁴,⁷.0⁵,³⁵.0²⁶,³⁴.0²⁷,³²]heptatriaconta-1,3,5(35),9,19,21,25(36),26(34),28,30,32-undecaen-13-yl acetate
Synonyms: rifaximin 利福昔明 rifacol normix fatroximin ritacol xifaxan
Molecular Weight (g/mol): 785.89
Molecular Formula: C43H51N3O11
InChi Key: NZCRJKRKKOLAOJ-XRCRFVBUSA-N
SMILES: CO[C@H]1\C=C\O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C1=C(N=C5C=C(C)C=CN15)C4=C3C2=O
Molecular Weight (g/mol) 
  • (9)
  • (2)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (3)
  • (5)
  • (9)
  • (2)
  • (3)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)