accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 80789-69-1

IUPAC Name: 1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
Synonyms: 1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 224.68
Molecular Formula: C12H13ClO2
InChi Key: QJNFJEMGWIQMJT-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCCC1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results