accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSOOONNa

CAS RN 80789-76-0

IUPAC Name: sodium 8-ethoxyquinoline-5-sulfonate hydrate
Synonyms: sodium 8-ethoxyquinoline-5-sulfonate hydrate
Molecular Weight (g/mol): 293.27
Molecular Formula: C11H12NNaO5S
InChi Key: BSVDXMZRSQZXAH-UHFFFAOYSA-M
SMILES: O.[Na+].CCOC1=C2N=CC=CC2=C(C=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results