accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 80791-87-3

N(R)N(R)O(R)(R)NNONH2HOHOH3CH2N

CAS RN 80791-87-3

IUPAC Name: 5-(2,6-diamino-9H-purin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
Synonyms: 5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
Molecular Weight (g/mol): 296.29
Molecular Formula: C11H16N6O4
InChi Key: JLWUWXCKSOIFPS-UHFFFAOYNA-N
SMILES: COC1C(O)C(CO)OC1N1C=NC2=C(N)N=C(N)N=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results