accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOONH2NHN

CAS RN 80983-34-2

IUPAC Name: 6-(propane-1-sulfonyl)-1H-1,3-benzodiazol-2-amine
Synonyms: 5-(propane-1-sulfonyl)-3H-1,3-benzodiazol-2-amine
Molecular Weight (g/mol): 239.29
Molecular Formula: C10H13N3O2S
InChi Key: WTPBIYSMFKUQKY-UHFFFAOYSA-N
SMILES: CCCS(=O)(=O)C1=CC=C2N=C(N)NC2=C1
Percent Purity 
  • (2)
  • (3)
Certifications/Compliance 
  • (1)
  • (2)
Industry Type 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)