accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOO

CAS RN 81-30-1

IUPAC Name: 6,13-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
Synonyms: naphthalenetetracarboxylic dianhydride
Molecular Weight (g/mol): 268.18
Molecular Formula: C14H4O6
InChi Key: YTVNOVQHSGMMOV-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CC=C3C(=O)OC(=O)C4=CC=C1C2=C34
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results