accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COClHOOO

CAS RN 81-82-3

IUPAC Name: 3-[1-(4-chlorophenyl)-3-oxobutyl]-2-hydroxy-4H-chromen-4-one
Synonyms: coumachlor
Molecular Weight (g/mol): 342.78
Molecular Formula: C19H15ClO4
InChi Key: HENZOLWOIZODCT-UHFFFAOYNA-N
SMILES: CC(=O)CC(C1=CC=C(Cl)C=C1)C1=C(O)OC2=CC=CC=C2C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results