accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 81012-89-7

IUPAC Name: naphthalen-1-yl phosphate
Synonyms: naphthalen-1-yl phosphate
Molecular Weight (g/mol): 222.14
Molecular Formula: C10H7O4P
InChi Key: YNXICDMQCQPQEW-UHFFFAOYSA-L
SMILES: [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (4)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results