accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClOO

CAS RN 81028-92-4

IUPAC Name: 3-(3,5-dichlorophenoxy)benzaldehyde
Synonyms: 3-(3,5-dichlorophenoxy)benzaldehyde
Molecular Weight (g/mol): 267.11
Molecular Formula: C13H8Cl2O2
InChi Key: BISWHYILBVQCRA-UHFFFAOYSA-N
SMILES: ClC1=CC(OC2=CC=CC(C=O)=C2)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results