accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NHNNO

CAS RN 81078-84-4

IUPAC Name: 1-{furo[3,2-c]pyridin-4-yl}piperazine
Synonyms: 1-{furo[3,2-c]pyridin-4-yl}piperazine
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: LUIZVTKJWSGSPA-UHFFFAOYSA-N
SMILES: C1CN(CCN1)C1=C2C=COC2=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results