accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 81290-20-2

IUPAC Name: tert-butyl(trifluoromethyl)silane
Synonyms: trifluoromethyltrimethylsilane
Molecular Weight (g/mol): 156.22
Molecular Formula: C5H11F3Si
InChi Key: MTYSDPUZDZURPP-UHFFFAOYSA-N
SMILES: CC(C)(C)[SiH2]C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (13)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (13)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (7)
  • (3)
  • (3)