accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 813-78-5

CH3OPOHOOH3C

CAS RN 813-78-5

IUPAC Name: dimethoxyphosphinic acid
Synonyms: dimethyl phosphate
Molecular Weight (g/mol): 126.05
Molecular Formula: C2H7O4P
InChi Key: KKUKTXOBAWVSHC-UHFFFAOYSA-N
SMILES: COP(O)(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)