accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 814-68-6

IUPAC Name: prop-2-enoyl chloride
Synonyms: acryloyl chloride
Molecular Weight (g/mol): 90.51
Molecular Formula: C3H3ClO
InChi Key: HFBMWMNUJJDEQZ-UHFFFAOYSA-N
SMILES: ClC(=O)C=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results