accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SHO

CAS RN 81443-44-9

IUPAC Name: [4-(thiophen-2-yl)phenyl]methanol
Synonyms: [4-(thiophen-2-yl)phenyl]methanol
Molecular Weight (g/mol): 190.26
Molecular Formula: C11H10OS
InChi Key: KHZSWSHUIYOMSH-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results