CAS RN 816-57-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitroso-N-propyl Urea, Contains 40% Water, ~2% Acetic Acid, TRC
CAS: 816-57-9 Molecular Formula: C4 H9 N3 O2 Molecular Weight (g/mol): 131.13 Synonym: Urea, N-nitroso-N-propyl- (9CI, ACI),N-Nitroso-N-propylurea (ACI),Urea, 1-nitroso-1-propyl- (7CI, 8CI),1-n-Propyl-1-nitrosourea,1-Nitroso-1-propylurea,1-Propyl-1-nitrosourea,N-(n-Propyl)-N-nitrosourea,N-Propyl-N-nitrosourea,NPU,NSC 46792 IUPAC Name: 1-nitroso-1-propylurea SMILES: CCCN(N=O)C(=O)N