accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 81661-26-9

CH3OOOOCH3

CAS RN 81661-26-9

IUPAC Name: methyl 5-formyl-2-methylfuran-3-carboxylate
Synonyms: methyl 5-formyl-2-methylfuran-3-carboxylate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: WINMHOHYPKKTEM-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)OC(C=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results